Gene Transcripts

Gene Symbol
Prr7
Entrez Gene ID
498704
Gene Name
proline rich 7 (synaptic)
Gene Aliases
Chromosome Location
Chr. 17: 9694157 - 9702438 on Build Rnor_6.0
UniGene ID
Rn.162185
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109116.1 NP_001102586.1 2 526
XM_006253652.2 XP_006253714.1 2 770

View Full Product Details