Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005683.3 | NP_005674.2 | 2 | 659 | |
XM_005246952.3 | XP_005247009.1 | 2 | 919 | ||
XM_011512175.2 | XP_011510477.1 | 2 | 1095 | ||
XM_011512176.2 | XP_011510478.1 | 2 | 678 | ||
GenBank mRNA | AK292223.1 | - | 2 | 804 | |
BC032694.1 | - | 2 | 659 | 5228678 | |
CR541776.1 | - | 1 | 466 | ||
FJ348259.1 | - | 1 | 466 |