Gene Transcripts

Gene Symbol
Xrcc2
Entrez Gene ID
499966
Gene Name
X-ray repair complementing defective repair in Chinese hamster cells 2
Gene Aliases
RGD1564823
Chromosome Location
Chr. 4: 5842013 - 5860516 on Build Rnor_6.0
UniGene ID
Rn.8224
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109215.1 NP_001102685.1 3 849
GenBank mRNA FQ211933.1 - 3 880

View Full Product Details