Gene Transcripts

Gene Symbol
Fmc1
Entrez Gene ID
500087
Gene Name
formation of mitochondrial complexes 1 homolog (S. cerevisiae)
Gene Aliases
Chromosome Location
Chr. 4: 66276835 - 66284868 on Build Rnor_6.0
UniGene ID
Rn.65180
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037795.1 NP_001032884.1 1 157
GenBank mRNA BC098843.1 - 1 157

View Full Product Details