Gene Transcripts

Gene Symbol
Tmco4
Entrez Gene ID
500573
Gene Name
transmembrane and coiled-coil domains 4
Gene Aliases
Chromosome Location
Chr. 5: 157421740 - 157501107 on Build Rnor_6.0
UniGene ID
Rn.162838
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034949.1 NP_001030121.1 9, 10 1102
XM_006239262.1 XP_006239324.1 9, 10 2527
GenBank mRNA BC099755.1 - 9, 10 1102

View Full Product Details