Gene Transcripts

Gene Symbol
STOML1
Entrez Gene ID
9399
Gene Name
stomatin like 1
Gene Aliases
SLP-1, STORP, hUNC-24
Chromosome Location
Chr. 15: 73983218 - 73994622 on Build GRCh38
UniGene ID
Hs.194816
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256672.1 NP_001243601.1 3 446
NM_001256675.1 NP_001243604.1 3 446
NM_001256676.1 NP_001243605.1 3 260
NM_001256677.1 NP_001243606.1 4 577
NM_001324226.1 NP_001311155.1 3 260
NM_001324227.1 NP_001311156.1 3 260
NM_001324228.1 NP_001311157.1 3 324
NM_001324229.1 NP_001311158.1 3 446
NM_001324230.1 NP_001311159.1 4 577
NM_004809.4 NP_004800.2 3 446
XM_005254792.4 XP_005254849.1 3 1350
XM_006720773.3 XP_006720836.1 3 1351
GenBank mRNA AF074953.1 - 3 308
AF156557.1 - 3 454
AK075244.1 - 3 342
AK091531.1 - 3 246
AK222665.1 - 3 329
AK300799.1 - 4 461
AL109665.1 - 3 300
AL533460.3 - 3 237
BC034379.1 - 3 392 5185908
BC065249.1 - 3 392 6059393
BX464941.2 - 3 260
CR542089.1 - 3 321
DQ064605.1 - 3 321
Y16522.1 - 3 365

View Full Product Details