Gene Transcripts

Gene Symbol
Trappc2
Entrez Gene ID
501550
Gene Name
trafficking protein particle complex 2
Gene Aliases
RGD1306925
Chromosome Location
Chr. X: 29550868 - 29562135 on Build Rnor_6.0
UniGene ID
Rn.234340
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024965.1 NP_001020136.1 6 463
XM_006256857.1 XP_006256919.1 6 553

View Full Product Details