Gene Transcripts

Gene Symbol
Gspt2
Entrez Gene ID
501582
Gene Name
G1 to S phase transition 2
Gene Aliases
RGD1563213
Chromosome Location
Chr. X: 63959131 - 63961652 on Build Rnor_6.0
UniGene ID
Rn.45722
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109319.1 NP_001102789.1 1 938

View Full Product Details