Gene Transcripts

Gene Symbol
Or51t1
Entrez Gene ID
502357
Gene Name
olfactory receptor, family 51, subfamily T, member 1
Gene Aliases
RGD1563481
Chromosome Location
Chr. 1: 168084954 - 168093181 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008759843.1 XP_008758065.1 3 840

View Full Product Details