Gene Transcripts

Gene Symbol
Iqcf6
Entrez Gene ID
503229
Gene Name
IQ motif containing F6
Gene Aliases
RGD1562645
Chromosome Location
Chr. 8: 115328535 - 115329546 on Build Rnor_6.0
UniGene ID
Rn.141073
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109362.1 NP_001102832.1 2 217

View Full Product Details