Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001109368.1 | NP_001102838.1 | 2 | 675 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_003752110.3 | XP_003752158.1 | 7 | 1186 | |
XM_008773459.1 | XP_008771681.1 | 8 | 1114 |