Gene Transcripts
- Gene Symbol
- PAGE4
- Entrez Gene ID
- 9506
- Gene Name
- PAGE family member 4
- Gene Aliases
- CT16.7, GAGE-9, GAGEC1, JM-27, JM27, PAGE-1, PAGE-4
- Chromosome Location
- Chr. X: 49829260 - 49834264 on Build GRCh38
- UniGene ID
- Hs.441038
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001318877.1 |
NP_001305806.1
|
3, 4 |
234 |
|
NM_007003.4 |
NP_008934.1
|
3, 4 |
246 |
|
GenBank mRNA |
AF275258.1 |
- |
3, 4 |
232 |
|
AJ005894.1 |
- |
3, 4 |
230 |
|
AK312039.1 |
- |
3, 4 |
243 |
|
AL548759.3 |
- |
2, 3 |
197 |
|
BC010897.2 |
- |
2, 3 |
196 |
4043535 |
BG435968.1 |
- |
3, 4 |
247 |
|
BG622311.1 |
- |
|
248 |
|
CR456823.1 |
- |
2, 3 |
167 |
|
CR542183.1 |
- |
2, 3 |
167 |
|
DA841276.1 |
- |
3, 4 |
228 |
|
View Full Product Details