Gene Transcripts

Gene Symbol
Dmrtc1b
Entrez Gene ID
680068
Gene Name
DMRT-like family C1b
Gene Aliases
Chromosome Location
Chr. X: 73663862 - 73686316 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001055549.5 XP_001055549.2 5 591
XM_006257140.2 XP_006257202.1 6 636
XM_008773329.1 XP_008771551.1 5 3197

View Full Product Details