Gene Transcripts

Gene Symbol
Dhrs2
Entrez Gene ID
691464
Gene Name
dehydrogenase/reductase (SDR family) member 2
Gene Aliases
Chromosome Location
Chr. 15: 33876994 - 33891564 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001078405.6 XP_001078405.4 8, 9 808
XM_008770715.1 XP_008768937.1 8, 9 824

View Full Product Details