Gene Transcripts

Gene Symbol
Tspan15
Entrez Gene ID
679462
Gene Name
tetraspanin 15
Gene Aliases
Chromosome Location
Chr. 20: 31812558 - 31857191 on Build Rnor_6.0
UniGene ID
Rn.216503
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001115032.1 NP_001108504.1 4 488
GenBank mRNA BC158654.1 - 4 488

View Full Product Details