Gene Transcripts

Gene Symbol
Vom2r42
Entrez Gene ID
679557
Gene Name
vomeronasal 2 receptor, 42
Gene Aliases
Chromosome Location
Chr. 1: 149132320 - 149175214 on Build Rnor_6.0
UniGene ID
Rn.217635
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099511.1 NP_001092981.1 3 868

View Full Product Details