Gene Transcripts
- Gene Symbol
- MPDU1
- Entrez Gene ID
- 9526
- Gene Name
- mannose-P-dolichol utilization defect 1
- Gene Aliases
- CDGIF, HBEBP2BPA, Lec35, My008, PP3958, PQLC5, SL15
- Chromosome Location
- Chr. 17: 7583647 - 7588212 on Build GRCh38
- UniGene ID
- Hs.246381
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_004870.3 |
NP_004861.2
|
4, 5 |
598 |
|
NR_024603.1 |
|
4, 5 |
593 |
|
XM_006721598.2 |
XP_006721661.1
|
4, 5 |
397 |
|
XM_011524081.2 |
XP_011522383.1
|
4, 5 |
429 |
|
GenBank mRNA |
AF038961.1 |
- |
4, 5 |
398 |
|
AF059752.1 |
- |
4, 5 |
438 |
|
AK075299.1 |
- |
4, 5 |
397 |
|
AK225401.1 |
- |
4, 5 |
397 |
|
AK225432.1 |
- |
4, 5 |
393 |
|
AK297001.1 |
- |
4, 5 |
393 |
|
AK300140.1 |
- |
4, 5 |
384 |
|
AK314527.1 |
- |
4, 5 |
393 |
|
BC001898.1 |
- |
4, 5 |
409 |
3536216 |
DQ499597.1 |
- |
4, 5 |
382 |
|
KU178478.1 |
- |
4, 5 |
382 |
|
View Full Product Details