Gene Transcripts

Gene Symbol
Glipr2
Entrez Gene ID
679819
Gene Name
GLI pathogenesis-related 2
Gene Aliases
Chromosome Location
Chr. 5: 59415407 - 59446732 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001054584.5 XP_001054584.2 2 201
XM_006238140.2 XP_006238202.1 3 1029

View Full Product Details