Gene Transcripts

Gene Symbol
Prr22
Entrez Gene ID
680177
Gene Name
proline rich 22
Gene Aliases
Chromosome Location
Chr. 9: 10286755 - 10289326 on Build Rnor_6.0
UniGene ID
Rn.159566
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001126375.1 NP_001119847.2 2 1268
XM_006244338.2 XP_006244400.1 3 1249
GenBank mRNA BC161872.1 - 2 1268

View Full Product Details