Gene Transcripts

Gene Symbol
Prss47
Entrez Gene ID
680378
Gene Name
protease, serine, 47
Gene Aliases
Chromosome Location
Chr. 17: 2814264 - 2828355 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006253500.2 XP_006253562.1 2, 3 786
XM_008771453.1 XP_008769675.1 2, 3 787
XM_008771454.1 XP_008769676.1 2, 3 509

View Full Product Details