Gene Transcripts

Gene Symbol
Pou5f2
Entrez Gene ID
680620
Gene Name
POU domain class 5, transcription factor 2
Gene Aliases
Sprm-1, Sprm1
Chromosome Location
Chr. 2: 5437067 - 5438301 on Build Rnor_6.0
UniGene ID
Rn.109178
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081751.1 NP_001075220.1 1 286
GenBank mRNA L23864.1 - 1 286

View Full Product Details