Gene Transcripts

Gene Symbol
Tex28
Entrez Gene ID
690282
Gene Name
testis expressed 28
Gene Aliases
Chromosome Location
Chr. X: 156543891 - 156569470 on Build Rnor_6.0
UniGene ID
Rn.228208
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191103.1 NP_001178032.1 917
XM_008773612.1 XP_008771834.1 1022

View Full Product Details