Gene Transcripts

Gene Symbol
Fpr2
Entrez Gene ID
690158
Gene Name
formyl peptide receptor 2
Gene Aliases
Fpr-rs2, Fprl1
Chromosome Location
Chr. 1: 59801178 - 59809507 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001073508.4 XP_001073508.1 2 698

View Full Product Details