Gene Transcripts

Gene Symbol
Tmem41a
Entrez Gene ID
681708
Gene Name
transmembrane protein 41a
Gene Aliases
Chromosome Location
Chr. 11: 82734525 - 82741177 on Build Rnor_6.0
UniGene ID
Rn.199221
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100805.1 NP_001094275.1 2 289
GenBank mRNA AY313923.1 - 1 65
BC161997.1 - 2 289 9035693

View Full Product Details