Gene Transcripts

Gene Symbol
Gzmn
Entrez Gene ID
691668
Gene Name
granzyme N
Gene Aliases
Chromosome Location
Chr. 15: 35129622 - 35160290 on Build Rnor_6.0
UniGene ID
Rn.137882
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191116.1 NP_001178045.1 5 654
XM_008770701.1 XP_008768923.1 6 752

View Full Product Details