Gene Transcripts

Gene Symbol
Wdr64
Entrez Gene ID
689524
Gene Name
WD repeat domain 64
Gene Aliases
Chromosome Location
Chr. 13: 93751756 - 93874137 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006250343.2 XP_006250405.1 6 761
XM_008769808.1 XP_008768030.1 6 761

View Full Product Details