Gene Transcripts

Gene Symbol
Clrn2
Entrez Gene ID
689515
Gene Name
clarin 2
Gene Aliases
Chromosome Location
Chr. 14: 70150983 - 70160624 on Build Rnor_6.0
UniGene ID
Rn.228209
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191097.1 NP_001178026.1 1 162

View Full Product Details