Gene Transcripts

Gene Symbol
Slirp
Entrez Gene ID
688717
Gene Name
SRA stem-loop interacting RNA binding protein
Gene Aliases
Chromosome Location
Chr. 6: 111476768 - 111486039 on Build Rnor_6.0
UniGene ID
Rn.65910
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109507.1 NP_001102977.1 2 214
GenBank mRNA BC166913.1 - 2 125
FQ223545.1 - 2 125

View Full Product Details