Gene Transcripts

Gene Symbol
Lilrc2
Entrez Gene ID
690906
Gene Name
leukocyte immunoglobulin-like receptor, subfamily C, member 2
Gene Aliases
Chromosome Location
Chr. 1: 63679228 - 63684274 on Build Rnor_6.0
UniGene ID
Rn.144780
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100123.1 NP_001093593.1 3 231
XM_006228169.2 XP_006228231.1 2 278
XM_006228170.2 XP_006228232.1 3 316
GenBank mRNA AY998483.1 - 3 210

View Full Product Details