Gene Transcripts

Gene Symbol
Lpgat1
Entrez Gene ID
679692
Gene Name
lysophosphatidylglycerol acyltransferase 1
Gene Aliases
Chromosome Location
Chr. 13: 110397347 - 110470874 on Build Rnor_6.0
UniGene ID
Rn.225027
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109376.2 NP_001102846.1 1 273
XM_008769875.1 XP_008768097.1 2 331

View Full Product Details