Gene Transcripts

Gene Symbol
Clec2e
Entrez Gene ID
689853
Gene Name
C-type lectin domain family 2, member E
Gene Aliases
Clec2d10
Chromosome Location
Chr. 4: 162849880 - 162866382 on Build Rnor_6.0
UniGene ID
Rn.222006
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177689.1 NP_001171160.1 4 446
XM_008763336.1 XP_008761558.1 5 2538
GenBank mRNA FJ404473.1 - 4 482

View Full Product Details