Gene Transcripts

Gene Symbol
Skint4
Entrez Gene ID
313485
Gene Name
selection and upkeep of intraepithelial T cells 4
Gene Aliases
RGD1566327
Chromosome Location
Chr. 5: 132005656 - 132043403 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006238600.2 XP_006238662.2 4 694

View Full Product Details