Gene Transcripts

Gene Symbol
Snx3
Entrez Gene ID
684097
Gene Name
sorting nexin 3
Gene Aliases
Chromosome Location
Chr. 20: 47225382 - 47263390 on Build Rnor_6.0
UniGene ID
Rn.190405
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001044283.1 NP_001037748.1 3 631
GenBank mRNA BC086341.1 - 3 395
BC107918.1 - 3 631 7458016
BC158608.1 - 3 355 7301775
FQ211490.1 - 3 336
FQ231492.1 - 3 378
FQ234595.1 - 3 378

View Full Product Details