Gene Transcripts

Gene Symbol
Eif4e1b
Entrez Gene ID
686104
Gene Name
eukaryotic translation initiation factor 4E family member 1B
Gene Aliases
Chromosome Location
Chr. 17: 10369153 - 10372912 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008771512.1 XP_008769734.1 6 388

View Full Product Details