Gene Transcripts

Gene Symbol
Vom2r24
Entrez Gene ID
690816
Gene Name
vomeronasal 2 receptor, 24
Gene Aliases
Chromosome Location
Chr. 8: 3973634 - 4003593 on Build Rnor_6.0
UniGene ID
Rn.217947
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099493.1 NP_001092963.1 5, 6 1664

View Full Product Details