Gene Transcripts

Gene Symbol
Ankrd13c
Entrez Gene ID
685374
Gene Name
ankyrin repeat domain 13C
Gene Aliases
Chromosome Location
Chr. 2: 264785380 - 264833283 on Build Rnor_6.0
UniGene ID
Rn.100818
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191570.1 NP_001178499.1 9 1196
XM_006233547.2 XP_006233609.1 8 2121
XM_008761571.1 XP_008759793.1 7 2017

Gene Symbol
LOC102548397
Entrez Gene ID
102548397
Gene Name
ankyrin repeat domain-containing protein 13C-like
Gene Aliases
Chromosome Location
Chr. 5: 132660535 - 132662584 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008763971.1 XP_008762193.1 1 1153

View Full Product Details