Gene Transcripts

Gene Symbol
Emid1
Entrez Gene ID
685462
Gene Name
EMI domain containing 1
Gene Aliases
RGD1565846
Chromosome Location
Chr. 14: 85357221 - 85400407 on Build Rnor_6.0
UniGene ID
Rn.81507
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109467.1 NP_001102937.1 7 848
XM_008770368.1 XP_008768590.1 14 1219

View Full Product Details