Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014728.3 | NP_055543.2 | 2 | 653 | |
XM_005274632.3 | XP_005274689.2 | 2 | 331 | ||
XM_011545613.2 | XP_011543915.1 | 2 | 640 | ||
XM_017029983.1 | XP_016885472.1 | 2 | 543 | ||
XM_017029984.1 | XP_016885473.1 | 2 | 332 | ||
XM_017029985.1 | XP_016885474.1 | 2 | 329 | ||
XM_017029986.1 | XP_016885475.1 | 2 | 542 | ||
GenBank mRNA | AB002314.3 | - | 2 | 653 | |
AK289694.1 | - | 2 | 640 | ||
BC113700.1 | - | 2 | 238 | 8322752 | |
BC113702.1 | - | 2 | 238 | 8322754 | |
BC143746.1 | - | 3 | 335 | 9052260 |