Gene Transcripts

Gene Symbol
Znrd1-as1
Entrez Gene ID
685722
Gene Name
ZNRD1 antisense RNA 1
Gene Aliases
Chromosome Location
Chr. 9: 18489284 - 18490450 on Build Rnor_6.0
UniGene ID
Rn.136265
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109479.1 NP_001102949.1 1 678

View Full Product Details