Gene Transcripts

Gene Symbol
Sh2d1b2
Entrez Gene ID
685844
Gene Name
SH2 domain containing 1B2
Gene Aliases
Chromosome Location
Chr. 13: 88557320 - 88581367 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001065477.4 XP_001065477.2 2, 3 863

View Full Product Details