Gene Transcripts

Gene Symbol
Ifnl3
Entrez Gene ID
685840
Gene Name
interferon, lambda 3
Gene Aliases
Il28b
Chromosome Location
Chr. 1: 85236243 - 85237775 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006228638.1 XP_006228700.1 1 117

Gene Symbol
LOC100911249
Entrez Gene ID
100911249
Gene Name
interleukin-28A-like
Gene Aliases
Chromosome Location
Chr. 1: 85252103 - 85253635 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008759287.1 XP_008757509.1 1 117

View Full Product Details