Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014740.3 | NP_055555.1 | 7, 8 | 943 | |
XM_011525522.1 | XP_011523824.1 | 6, 7 | 836 | ||
GenBank mRNA | AK290608.1 | - | 7, 8 | 924 | |
BC003662.2 | - | 7, 8 | 853 | 2988931 | |
BC004386.1 | - | 7, 8 | 874 | 3619386 | |
BC011151.1 | - | 7, 8 | 878 | 3452439 | |
CR456750.1 | - | 7, 8 | 721 | ||
D21853.1 | - | 7, 8 | 935 | ||
X79538.1 | - | 7, 8 | 794 |