Gene Transcripts

Gene Symbol
Fam47e
Entrez Gene ID
686140
Gene Name
family with sequence similarity 47, member E
Gene Aliases
Fam47e-ps1
Chromosome Location
Chr. 14: 16991496 - 17027568 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008770095.1 XP_008768317.1 9 1311
XM_008770096.1 XP_008768318.1 8 1218

View Full Product Details