Gene Transcripts

Gene Symbol
Smg5
Entrez Gene ID
681012
Gene Name
SMG5 nonsense mediated mRNA decay factor
Gene Aliases
Chromosome Location
Chr. 2: 187707331 - 187735355 on Build Rnor_6.0
UniGene ID
Rn.203043
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001059906.5 XP_001059906.3 1 1093
GenBank mRNA BC169031.1 - 1 151

View Full Product Details