Gene Transcripts

Gene Symbol
Iqsec1
Entrez Gene ID
686590
Gene Name
IQ motif and Sec7 domain 1
Gene Aliases
Chromosome Location
Chr. 4: 123400730 - 123454656 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006236890.2 XP_006236952.1 3 1519
XM_008763140.1 XP_008761362.1 3 1519
XM_008763141.1 XP_008761363.1 3 1519

View Full Product Details