Gene Transcripts

Gene Symbol
Ckap2
Entrez Gene ID
306575
Gene Name
cytoskeleton associated protein 2
Gene Aliases
Chromosome Location
Chr. 16: 74752655 - 74779184 on Build Rnor_6.0
UniGene ID
Rn.50526
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001169139.1 NP_001162610.1 2 141
GenBank mRNA EV774137.1 - 2 123

View Full Product Details