Gene Transcripts

Gene Symbol
Vom2r55
Entrez Gene ID
691442
Gene Name
vomeronasal 2 receptor, 55
Gene Aliases
Chromosome Location
Chr. 7: 16595532 - 16623355 on Build Rnor_6.0
UniGene ID
Rn.217934
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099495.1 NP_001092965.1 1915

View Full Product Details