Gene Transcripts

Gene Symbol
Mamstr
Entrez Gene ID
691041
Gene Name
MEF2 activating motif and SAP domain containing transcriptional regulator
Gene Aliases
Chromosome Location
Chr. 1: 101621704 - 101628379 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001076631.5 XP_001076631.2 5 1360
XM_006229210.2 XP_006229272.1 5 1745
XM_006229211.2 XP_006229273.1 5 1304

View Full Product Details