Gene Transcripts
- Gene Symbol
- Rpl22l1
- Entrez Gene ID
- 361923
- Gene Name
- ribosomal protein L22 like 1
- Gene Aliases
- RGD1309517
- Chromosome Location
- Chr. 2: 115362799 - 115364735 on Build Rnor_6.0
- UniGene ID
- Rn.112006
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001108548.1 |
NP_001102018.1
|
2, 3 |
207 |
|
GenBank mRNA |
BC167114.1 |
- |
2, 3 |
124 |
|
FQ210535.1 |
- |
2, 3 |
123 |
|
FQ211167.1 |
- |
2, 3 |
123 |
|
FQ221224.1 |
- |
2, 3 |
123 |
|
FQ221258.1 |
- |
2, 3 |
121 |
|
FQ221490.1 |
- |
2, 3 |
123 |
|
FQ221726.1 |
- |
2, 3 |
123 |
|
FQ221812.1 |
- |
2, 3 |
123 |
|
FQ221904.1 |
- |
2, 3 |
124 |
|
FQ223163.1 |
- |
2, 3 |
123 |
|
FQ223463.1 |
- |
2, 3 |
123 |
|
FQ223753.1 |
- |
2, 3 |
122 |
|
FQ228314.1 |
- |
2, 3 |
123 |
|
FQ228345.1 |
- |
2, 3 |
123 |
|
FQ229046.1 |
- |
2, 3 |
123 |
|
View Full Product Details