Gene Transcripts

Gene Symbol
Vps25
Entrez Gene ID
681059
Gene Name
vacuolar protein sorting 25 homolog (S. cerevisiae)
Gene Aliases
Chromosome Location
Chr. 10: 89174684 - 89180165 on Build Rnor_6.0
UniGene ID
Rn.6985
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001173451.1 NP_001166922.1 4 296
GenBank mRNA FQ213477.1 - 4 276
FQ220187.1 - 4 276
FQ229682.1 - 4 276
FQ233075.1 - 4 277

View Full Product Details